Product Details

SNP ID
rs201761312
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:15485946 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCTGAAGCAGGGCCAGTTGCTGG[G/T]TGTGGACTATGCCACCTGCTCCAGC
Phenotype
MIM: 602234 MIM: 609444
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CASP9 PubMed Links

Gene Details

Gene
CASP9
Gene Name
caspase 9
There are no transcripts associated with this gene.

Gene
CELA2B
Gene Name
chymotrypsin like elastase family member 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015849.2 564 Missense Mutation GGT,GTT G180V NP_056933.2

View Full Product Details