Product Details

SNP ID
rs201626227
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:23089010 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGACGGATCCAGGGAGTTGTGC[A/G]GTTGCCTGTAGACACTGAGTGTACA
Phenotype
MIM: 609132 MIM: 601422
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KDM1A PubMed Links

Gene Details

Gene
KDM1A
Gene Name
lysine demethylase 1A
There are no transcripts associated with this gene.

Gene
LUZP1
Gene Name
leucine zipper protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142546.1 3500 Missense Mutation CCG,CTG P1039L NP_001136018.1
NM_033631.3 3500 Missense Mutation CCG,CTG P1039L NP_361013.3
XM_011542090.2 3500 Missense Mutation CCG,CTG P1039L XP_011540392.1
XM_011542091.2 3500 Missense Mutation CCG,CTG P1039L XP_011540393.1
XM_017002252.1 3500 Missense Mutation CCG,CTG P1039L XP_016857741.1

View Full Product Details