Product Details

SNP ID
rs201152024
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:115286240 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTCAATGCCCCGGCACCCGCTGT[C/T]AACGGGATTTGGGTCCCGGCACTTG
Phenotype
MIM: 162030
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NGF PubMed Links

Gene Details

Gene
NGF
Gene Name
nerve growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002506.2 725 Missense Mutation AAC,GAC N186D NP_002497.2
XM_006710663.3 725 Missense Mutation AAC,GAC N186D XP_006710726.1
XM_011541518.2 725 Missense Mutation AAC,GAC N241D XP_011539820.1

View Full Product Details