Product Details

SNP ID
rs201575101
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:151158807 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTTCATAGATGAGACAAGCAGTG[A/C]GGTGCTAGATGAGCTCTACCGTGTG
Phenotype
MIM: 608095 MIM: 612112
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LYSMD1 PubMed Links

Gene Details

Gene
LYSMD1
Gene Name
LysM domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136543.1 236 Intron NP_001130015.1
NM_212551.4 236 Intron NP_997716.1
Gene
SCNM1
Gene Name
sodium channel modifier 1
There are no transcripts associated with this gene.

Gene
TNFAIP8L2
Gene Name
TNF alpha induced protein 8 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024575.4 236 Missense Mutation GAG,GCG E37A NP_078851.2
Gene
TNFAIP8L2-SCNM1
Gene Name
TNFAIP8L2-SCNM1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204848.1 236 Intron NP_001191777.1

View Full Product Details