Product Details

SNP ID
rs201641354
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:158420275 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAATGTACAGAGCATGAAGATGA[C/G]AATCTGACTAAAGTGGTTATGGTGA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR10K2 PubMed Links

Gene Details

Gene
OR10K2
Gene Name
olfactory receptor family 10 subfamily K member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004476.1 592 Missense Mutation CTC,GTC L198V NP_001004476.1

View Full Product Details