Product Details

SNP ID
rs201006238
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:231335986 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTTCATCTCTTCTTTGAGCTTA[C/T]CCAGGGCTTCTGGCGTCATCAAGTT
Phenotype
MIM: 615283 MIM: 616086
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXOC8 PubMed Links

Gene Details

Gene
EXOC8
Gene Name
exocyst complex component 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175876.4 1887 Missense Mutation GAT,GGT D587G NP_787072.2
Gene
SPRTN
Gene Name
SprT-like N-terminal domain
There are no transcripts associated with this gene.

View Full Product Details