Product Details

SNP ID
rs201521847
Assay Type
Functionally Tested
NCBI dbSNP Submissions
9
Location
Chr.1:152511824 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGTGGCAGTGGCCAGCAGTCTG[A/G]GGGCTCCAGCTGCTGCCACAGCTCT
Phenotype
MIM: 612619
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CRCT1 PubMed Links

Gene Details

Gene
CRCT1
Gene Name
cysteine rich C-terminal 1
There are no transcripts associated with this gene.

Gene
LCE5A
Gene Name
late cornified envelope 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178438.4 466 Missense Mutation GAG,GGG E97G NP_848525.1

View Full Product Details