Product Details

SNP ID
rs201694229
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:24745585 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTCGATGCCGCTGAATGGGCTGA[A/G]GGAGGAGGACAAAGAGCCCCTCATC
Phenotype
MIM: 606536
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLIC4 PubMed Links

Gene Details

Gene
CLIC4
Gene Name
chloride intracellular channel 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013943.2 317 Missense Mutation AAG,AGG K11R NP_039234.1

View Full Product Details