Product Details

SNP ID
rs201220123
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:224183363 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGAGCCGCGTCTCGCGGGAAGA[A/C]TTCGAGTGGGTCTACACCGACCAGC
Phenotype
MIM: 615843
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DEGS1 PubMed Links

Gene Details

Gene
DEGS1
Gene Name
delta 4-desaturase, sphingolipid 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321541.1 156 Missense Mutation GAA,GAC E9D NP_001308470.1
NM_001321542.1 156 Intron NP_001308471.1
NM_003676.3 156 Missense Mutation GAA,GAC E9D NP_003667.1
XM_017002648.1 156 Intron XP_016858137.1

View Full Product Details