Product Details

SNP ID
rs201233416
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:156244834 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAAGAGGAAGCCAGTGAGCAGCGC[A/G]CAGAAGAGATGGTAGCCATGGCTGG
Phenotype
MIM: 112260 MIM: 614579 MIM: 609176 MIM: 610962
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BGLAP PubMed Links

Gene Details

Gene
BGLAP
Gene Name
bone gamma-carboxyglutamate protein
There are no transcripts associated with this gene.

Gene
PAQR6
Gene Name
progestin and adipoQ receptor family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272104.1 915 Silent Mutation TGC,TGT C229C NP_001259033.1
NM_001272105.1 915 Silent Mutation TGC,TGT C226C NP_001259034.1
NM_001272106.1 915 Intron NP_001259035.1
NM_001272107.1 915 Silent Mutation TGC,TGT C123C NP_001259036.1
NM_001272108.1 915 Silent Mutation TGC,TGT C118C NP_001259037.1
NM_001272109.1 915 Missense Mutation GCG,GTG A7V NP_001259038.1
NM_001272110.1 915 Missense Mutation GCG,GTG A7V NP_001259039.1
NM_001272111.1 915 Missense Mutation GCG,GTG A7V NP_001259040.1
NM_001272112.1 915 Missense Mutation GCG,GTG A7V NP_001259041.1
NM_001272113.1 915 Missense Mutation GCG,GTG A7V NP_001259042.1
NM_024897.3 915 Silent Mutation TGC,TGT C123C NP_079173.2
NM_198406.2 915 Silent Mutation TGC,TGT C229C NP_940798.1
XM_005245494.3 915 Missense Mutation GCG,GTG A7V XP_005245551.1
XM_006711546.2 915 Silent Mutation TGC,TGT C229C XP_006711609.1
XM_006711547.3 915 Silent Mutation TGC,TGT C229C XP_006711610.1
XM_006711548.3 915 Silent Mutation TGC,TGT C205C XP_006711611.1
XM_006711552.2 915 Silent Mutation TGC,TGT C118C XP_006711615.1
XM_006711553.3 915 Silent Mutation TGC,TGT C118C XP_006711616.1
XM_011510000.2 915 Missense Mutation GCG,GTG A7V XP_011508302.1
XM_017002385.1 915 Silent Mutation TGC,TGT C229C XP_016857874.1
Gene
PMF1
Gene Name
polyamine-modulated factor 1
There are no transcripts associated with this gene.

Gene
PMF1-BGLAP
Gene Name
PMF1-BGLAP readthrough
There are no transcripts associated with this gene.

Gene
SMG5
Gene Name
SMG5, nonsense mediated mRNA decay factor
There are no transcripts associated with this gene.

View Full Product Details