Product Details

SNP ID
rs201764848
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:110061514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTTCCGCCCCTCCTGGATCTTC[C/G]CATAACGCTCGCGCTTCCGCCACTT
Phenotype
MIM: 606014
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ALX3 PubMed Links

Gene Details

Gene
ALX3
Gene Name
ALX homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006492.2 732 Missense Mutation GCG,GGG A215G NP_006483.2
Gene
STRIP1
Gene Name
striatin interacting protein 1
There are no transcripts associated with this gene.

View Full Product Details