Product Details

SNP ID
rs201630556
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:54609605 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGGCCAGCCTGGTGCCACAGTC[A/T]CGTGGGGGAAGACCTCAGCCACAGC
Phenotype
MIM: 606803
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ACOT11 PubMed Links

Gene Details

Gene
ACOT11
Gene Name
acyl-CoA thioesterase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015547.3 1629 Intron NP_056362.1
NM_147161.3 1629 UTR 3 NP_671517.1
Gene
FAM151A
Gene Name
family with sequence similarity 151 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176782.2 1629 Intron NP_788954.2
XM_006710599.3 1629 Missense Mutation GAG,GTG E448V XP_006710662.1

View Full Product Details