Product Details

SNP ID
rs201696505
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:155054322 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTCTTTTTTCTTAGAGGCTTGG[G/T]GGTCCTGACCCCAGAGAGAAGCTAT
Phenotype
MIM: 605548 MIM: 601380
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ADAM15 PubMed Links

Gene Details

Gene
ADAM15
Gene Name
ADAM metallopeptidase domain 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261464.1 543 Missense Mutation GGG,GTG G153V NP_001248393.1
NM_001261465.1 543 Missense Mutation GGG,GTG G143V NP_001248394.1
NM_001261466.1 543 Missense Mutation GGG,GTG G127V NP_001248395.1
NM_003815.4 543 Missense Mutation GGG,GTG G143V NP_003806.3
NM_207191.2 543 Missense Mutation GGG,GTG G143V NP_997074.1
NM_207194.2 543 Missense Mutation GGG,GTG G143V NP_997077.1
NM_207195.2 543 Missense Mutation GGG,GTG G143V NP_997078.1
NM_207196.2 543 Missense Mutation GGG,GTG G143V NP_997079.1
NM_207197.2 543 Missense Mutation GGG,GTG G143V NP_997080.1
Gene
DCST1
Gene Name
DC-STAMP domain containing 1
There are no transcripts associated with this gene.

Gene
EFNA4
Gene Name
ephrin A4
There are no transcripts associated with this gene.

Gene
LOC100505666
Gene Name
uncharacterized LOC100505666
There are no transcripts associated with this gene.

View Full Product Details