Product Details

SNP ID
rs201262598
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:43653249 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTATCCAACTATGGAAGAGTTCC[A/G]AAACTTCAGTAGATACATTGCCTAC
Phenotype
MIM: 609764
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KDM4A PubMed Links

Gene Details

Gene
KDM4A
Gene Name
lysine demethylase 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014663.2 277 Missense Mutation CAA,CGA Q25R NP_055478.2
XM_005271354.3 277 Missense Mutation CAA,CGA Q25R XP_005271411.1
XM_005271355.3 277 Missense Mutation CAA,CGA Q25R XP_005271412.1
XM_005271356.3 277 UTR 5 XP_005271413.1
XM_011542467.2 277 Intron XP_011540769.2
XM_017002912.1 277 UTR 5 XP_016858401.1

View Full Product Details