Product Details

SNP ID
rs201618935
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:1327477 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCGCGCCCTGCACTGGCTGCAGC[C/T]GTTCCTGGAGGGCCTGCGTACCAGC
Phenotype
MIM: 611354 MIM: 615467 MIM: 601365 MIM: 605865
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPSF3L PubMed Links

Gene Details

Gene
CPSF3L
Gene Name
cleavage and polyadenylation specific factor 3-like
There are no transcripts associated with this gene.

Gene
CPTP
Gene Name
ceramide-1-phosphate transfer protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029885.1 774 Missense Mutation CCG,CTG P120L NP_001025056.1
XM_005244801.3 774 Missense Mutation CCG,CTG P120L XP_005244858.1
XM_005244802.1 774 Missense Mutation CCG,CTG P57L XP_005244859.1
XM_011542200.2 774 Missense Mutation CCG,CTG P120L XP_011540502.1
Gene
DVL1
Gene Name
dishevelled segment polarity protein 1
There are no transcripts associated with this gene.

Gene
TAS1R3
Gene Name
taste 1 receptor member 3
There are no transcripts associated with this gene.

View Full Product Details