Product Details

SNP ID
rs201659515
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:45341507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAACGTTACAAGGCCGTGTGTCAT[C/G]CTGCCAGGACCCGTTCTATCCTTTC
Phenotype
MIM: 604933 MIM: 604746 MIM: 613931
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MUTYH PubMed Links

Gene Details

Gene
MUTYH
Gene Name
mutY DNA glycosylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001048171.1 1114 Intron NP_001041636.1
NM_001048172.1 1114 Intron NP_001041637.1
NM_001048173.1 1114 Intron NP_001041638.1
NM_001048174.1 1114 Intron NP_001041639.1
NM_001128425.1 1114 Intron NP_001121897.1
NM_001293190.1 1114 Intron NP_001280119.1
NM_001293191.1 1114 Intron NP_001280120.1
NM_001293192.1 1114 Intron NP_001280121.1
NM_001293195.1 1114 Intron NP_001280124.1
NM_001293196.1 1114 Intron NP_001280125.1
NM_012222.2 1114 Intron NP_036354.1
XM_011541497.2 1114 Intron XP_011539799.1
XM_011541498.1 1114 Intron XP_011539800.1
XM_011541499.1 1114 Intron XP_011539801.1
XM_011541500.2 1114 Intron XP_011539802.1
XM_011541501.2 1114 Intron XP_011539803.1
XM_011541502.2 1114 Intron XP_011539804.1
XM_011541503.2 1114 Intron XP_011539805.1
XM_011541504.2 1114 Intron XP_011539806.1
XM_011541505.2 1114 Intron XP_011539807.1
XM_011541506.1 1114 Intron XP_011539808.1
XM_011541507.2 1114 Intron XP_011539809.2
XM_017001331.1 1114 Intron XP_016856820.1
XM_017001332.1 1114 Intron XP_016856821.1
XM_017001333.1 1114 Intron XP_016856822.1
XM_017001334.1 1114 Intron XP_016856823.1
XM_017001335.1 1114 Intron XP_016856824.1
XM_017001336.1 1114 Intron XP_016856825.1
XM_017001337.1 1114 Intron XP_016856826.1
XM_017001338.1 1114 Intron XP_016856827.1
XM_017001339.1 1114 Intron XP_016856828.1
Gene
TESK2
Gene Name
testis-specific kinase 2
There are no transcripts associated with this gene.

Gene
TOE1
Gene Name
target of EGR1, member 1 (nuclear)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025077.3 1114 Missense Mutation CCT,GCT P91A NP_079353.3
XM_005270412.3 1114 Missense Mutation CCT,GCT P97A XP_005270469.1
XM_005270413.4 1114 Missense Mutation CCT,GCT P45A XP_005270470.1
XM_011540569.2 1114 Intron XP_011538871.1

View Full Product Details