Product Details

SNP ID
rs201382965
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:150984058 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCATCTCACGAAACTTCCAGGAG[C/T]GCACCCAACAGGTGAGGCCATGCTG
Phenotype
MIM: 603319 MIM: 606920
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANXA9 PubMed Links

Gene Details

Gene
ANXA9
Gene Name
annexin A9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003568.2 445 Missense Mutation CGC,TGC R86C NP_003559.2
XM_011510058.2 445 Missense Mutation CGC,TGC R86C XP_011508360.1
Gene
CERS2
Gene Name
ceramide synthase 2
There are no transcripts associated with this gene.

View Full Product Details