Product Details

SNP ID
rs201037790
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:27330738 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACAGGTGGTTTTTCCCCATCAT[C/T]GGCCACATGGGCATCTGCACATCCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTG1P20 PubMed Links

Gene Details

Gene
ACTG1P20
Gene Name
actin gamma 1 pseudogene 20
There are no transcripts associated with this gene.

Gene
TMEM222
Gene Name
transmembrane protein 222
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032125.2 266 Silent Mutation ATC,ATT I71I NP_115501.2

View Full Product Details