Product Details

SNP ID
rs201515244
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:31791277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGGGAAAGGAGAGGGGAGACAG[A/G]AAGAGCCCAAAGGCCTGCGGGGAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPOCD1 PubMed Links

Gene Details

Gene
SPOCD1
Gene Name
SPOC domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281987.1 3107 Intron NP_001268916.1
NM_001281988.1 3107 Intron NP_001268917.1
NM_144569.5 3107 Missense Mutation NP_653170.3
XM_011542413.2 3107 Intron XP_011540715.1
XM_011542414.2 3107 Intron XP_011540716.1
XM_017002779.1 3107 Intron XP_016858268.1
XM_017002780.1 3107 Intron XP_016858269.1
XM_017002781.1 3107 Intron XP_016858270.1
XM_017002782.1 3107 Intron XP_016858271.1

View Full Product Details