Product Details

SNP ID
rs201741123
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:151364677 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGCCCTCCTTTTACTGTGTCTA[A/C]ATCAACCTGCAGCATCACAGAGCCT
Phenotype
MIM: 604188
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SELENBP1 PubMed Links

Gene Details

Gene
SELENBP1
Gene Name
selenium binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258288.1 1587 Missense Mutation GTA,TTA V367L NP_001245217.1
NM_001258289.1 1587 Missense Mutation GTA,TTA V471L NP_001245218.1
NM_003944.3 1587 Missense Mutation GTA,TTA V429L NP_003935.2
XM_017002757.1 1587 Intron XP_016858246.1

View Full Product Details