Product Details

SNP ID
rs201119972
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:54999275 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTACGTTCCTCATGAGCCATGATC[A/G]GCCCCAGGTCTACGGCACCTTCTAT
Phenotype
MIM: 606412
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BSND PubMed Links

Gene Details

Gene
BSND
Gene Name
barttin CLCNK type accessory beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_057176.2 332 Missense Mutation CAG,CGG Q30R NP_476517.1
Gene
TMEM61
Gene Name
transmembrane protein 61
There are no transcripts associated with this gene.

View Full Product Details