Product Details

SNP ID
rs201306033
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:159918850 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCCAGTCATGCCTGCCTGAGACG[C/T]CCCGCGGTTGGTGCCCATCTGTAAC
Phenotype
MIM: 609738 MIM: 604634
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGSF9 PubMed Links

Gene Details

Gene
IGSF9
Gene Name
immunoglobulin superfamily member 9
There are no transcripts associated with this gene.

Gene
TAGLN2
Gene Name
transgelin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277223.1 860 Intron NP_001264152.1
NM_001277224.1 860 Missense Mutation ACG,GCG T205A NP_001264153.1
NM_003564.2 860 Intron NP_003555.1

View Full Product Details