Product Details

SNP ID
rs201201896
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:246726996 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGCAACAGTTGTCCTCAATTGC[A/G]TAGGACCAGTAAGTAATCAACCCTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCCPDH PubMed Links

Gene Details

Gene
SCCPDH
Gene Name
saccharopine dehydrogenase (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016002.2 642 Missense Mutation ATA,GTA I99V NP_057086.2

View Full Product Details