Product Details

SNP ID
rs201730097
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:225985491 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCCAGTTCTGCAACAGAGGTGAA[C/T]GCCAATAAATCTATAGTTTCCTTAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SDE2 PubMed Links

Gene Details

Gene
SDE2
Gene Name
SDE2 telomere maintenance homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152608.3 1220 Silent Mutation GCA,GCG A389A NP_689821.3

View Full Product Details