Product Details

SNP ID
rs201055560
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:52412531 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGTAGAAACGCTATGTATTAGA[C/G]GAAGCTGAGCAACTGGAGCCTCGAG
Phenotype
MIM: 601902 MIM: 617031
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ORC1 PubMed Links

Gene Details

Gene
ORC1
Gene Name
origin recognition complex subunit 1
There are no transcripts associated with this gene.

Gene
PRPF38A
Gene Name
pre-mRNA processing factor 38A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032864.3 719 Missense Mutation GAC,GAG D172E NP_116253.2
XM_011542315.2 719 Missense Mutation GAC,GAG D172E XP_011540617.1

View Full Product Details