Product Details

SNP ID
rs201148660
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:150733130 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCCGAATATATCCTTCTTCACC[A/G]AAGTTGTGGCCCCAGCTTTAGAAAA
Phenotype
MIM: 116845
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTSS PubMed Links

Gene Details

Gene
CTSS
Gene Name
cathepsin S
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199739.1 1173 Silent Mutation TTC,TTT F254F NP_001186668.1
NM_004079.4 1173 Silent Mutation TTC,TTT F304F NP_004070.3

View Full Product Details