Product Details

SNP ID
rs201492707
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:168184869 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGAACCTTACTTGGAGAATCTCT[G/T]AAGTTAAAGGTAAATCTTACTTTTT
Phenotype
MIM: 611807
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TIPRL PubMed Links

Gene Details

Gene
TIPRL
Gene Name
TOR signaling pathway regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031800.2 608 Silent Mutation CTG,CTT L125L NP_001026970.1
NM_152902.4 608 Silent Mutation CTG,CTT L125L NP_690866.1

View Full Product Details