Product Details
- SNP ID
-
rs199609262
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
9
- Location
-
Chr.1:43995659 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACCCCTCTGAGGCAGCCCTGGAGC[C/G]CACCCTGGCAGGTGAGGACACGGAG
- Phenotype
-
MIM: 604013
MIM: 601019
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
B4GALT2
PubMed Links
Gene Details
- Gene
- B4GALT2
- Gene Name
- beta-1,4-galactosyltransferase 2
There are no transcripts associated with this gene.
- Gene
- CCDC24
- Gene Name
- coiled-coil domain containing 24
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152499.2 |
645 |
Missense Mutation |
CCC,CGC |
P204R |
NP_689712.1 |
XM_017000423.1 |
645 |
Missense Mutation |
CCC,CGC |
P285R |
XP_016855912.1 |
XM_017000424.1 |
645 |
Missense Mutation |
CCC,CGC |
P285R |
XP_016855913.1 |
XM_017000425.1 |
645 |
Missense Mutation |
CCC,CGC |
P285R |
XP_016855914.1 |
XM_017000426.1 |
645 |
Missense Mutation |
CCC,CGC |
P254R |
XP_016855915.1 |
XM_017000427.1 |
645 |
Missense Mutation |
CCC,CGC |
P253R |
XP_016855916.1 |
XM_017000428.1 |
645 |
Missense Mutation |
CCC,CGC |
P252R |
XP_016855917.1 |
XM_017000429.1 |
645 |
Missense Mutation |
CCC,CGC |
P249R |
XP_016855918.1 |
XM_017000430.1 |
645 |
Missense Mutation |
CCC,CGC |
P252R |
XP_016855919.1 |
XM_017000431.1 |
645 |
Missense Mutation |
CCC,CGC |
P216R |
XP_016855920.1 |
XM_017000432.1 |
645 |
Missense Mutation |
CCC,CGC |
P205R |
XP_016855921.1 |
XM_017000433.1 |
645 |
Missense Mutation |
CCC,CGC |
P204R |
XP_016855922.1 |
XM_017000434.1 |
645 |
Missense Mutation |
CCC,CGC |
P203R |
XP_016855923.1 |
XM_017000435.1 |
645 |
Missense Mutation |
CCC,CGC |
P200R |
XP_016855924.1 |
XM_017000436.1 |
645 |
Missense Mutation |
CCC,CGC |
P200R |
XP_016855925.1 |
XM_017000437.1 |
645 |
Missense Mutation |
CCC,CGC |
P186R |
XP_016855926.1 |
XM_017000438.1 |
645 |
Missense Mutation |
CCC,CGC |
P181R |
XP_016855927.1 |
XM_017000439.1 |
645 |
Missense Mutation |
CCC,CGC |
P173R |
XP_016855928.1 |
XM_017000440.1 |
645 |
Missense Mutation |
CCC,CGC |
P168R |
XP_016855929.1 |
XM_017000441.1 |
645 |
Missense Mutation |
CCC,CGC |
P168R |
XP_016855930.1 |
XM_017000442.1 |
645 |
Missense Mutation |
CCC,CGC |
P128R |
XP_016855931.1 |
XM_017000443.1 |
645 |
Intron |
|
|
XP_016855932.1 |
XM_017000444.1 |
645 |
Intron |
|
|
XP_016855933.1 |
XM_017000445.1 |
645 |
Missense Mutation |
CCC,CGC |
P71R |
XP_016855934.1 |
XM_017000446.1 |
645 |
Missense Mutation |
CCC,CGC |
P70R |
XP_016855935.1 |
- Gene
- SLC6A9
- Gene Name
- solute carrier family 6 member 9
View Full Product Details