Product Details

SNP ID
rs199716453
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:43653187 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAGGAGGGAAAATGGCTTCTGA[A/G]TCTGAAACTCTGAATCCCAGTGCTA
Phenotype
MIM: 609764
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KDM4A PubMed Links

Gene Details

Gene
KDM4A
Gene Name
lysine demethylase 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014663.2 215 Silent Mutation GAA,GAG E4E NP_055478.2
XM_005271354.3 215 Silent Mutation GAA,GAG E4E XP_005271411.1
XM_005271355.3 215 Silent Mutation GAA,GAG E4E XP_005271412.1
XM_005271356.3 215 UTR 5 XP_005271413.1
XM_011542467.2 215 Intron XP_011540769.2
XM_017002912.1 215 UTR 5 XP_016858401.1

View Full Product Details