Product Details
- SNP ID
-
rs199704407
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:40514741 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAAGCTTACAAAACTGGAAAAGAG[A/G]ATTGGATATATTATCACCCATGGAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EXO5
PubMed Links
Gene Details
- Gene
- EXO5
- Gene Name
- exonuclease 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022774.1 |
629 |
Missense Mutation |
GAA,GGA |
E66G |
NP_073611.1 |
XM_005271125.3 |
629 |
Missense Mutation |
GAA,GGA |
E66G |
XP_005271182.1 |
XM_005271126.3 |
629 |
Missense Mutation |
GAA,GGA |
E66G |
XP_005271183.1 |
XM_005271128.3 |
629 |
Missense Mutation |
GAA,GGA |
E66G |
XP_005271185.1 |
XM_011541970.2 |
629 |
Missense Mutation |
GAA,GGA |
E66G |
XP_011540272.1 |
XM_017002099.1 |
629 |
Missense Mutation |
GAA,GGA |
E66G |
XP_016857588.1 |
XM_017002100.1 |
629 |
Missense Mutation |
GAA,GGA |
E66G |
XP_016857589.1 |
XM_017002101.1 |
629 |
Missense Mutation |
GAA,GGA |
E66G |
XP_016857590.1 |
XM_017002102.1 |
629 |
Missense Mutation |
GAA,GGA |
E66G |
XP_016857591.1 |
XM_017002103.1 |
629 |
Missense Mutation |
GAA,GGA |
E66G |
XP_016857592.1 |
XM_017002104.1 |
629 |
Missense Mutation |
GAA,GGA |
E66G |
XP_016857593.1 |
View Full Product Details