Product Details

SNP ID
rs199743368
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:114570728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTTCTGAAGTTCCTTCTGTGCGT[G/T]TTCAATCATATGAACTAGATTTCTG
Phenotype
MIM: 605783
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BCAS2 PubMed Links

Gene Details

Gene
BCAS2
Gene Name
breast carcinoma amplified sequence 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005872.2 Intron NP_005863.1

View Full Product Details