Product Details

SNP ID
rs199891940
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:152985240 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAACACATTTGAAGCATGAATTCT[C/T]AGCAGCAGAAGCAGCCTTGCACCCC
Phenotype
MIM: 182265
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPRR1A PubMed Links

Gene Details

Gene
SPRR1A
Gene Name
small proline rich protein 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199828.1 67 Nonsense Mutation CAG,TAG Q4* NP_001186757.1
NM_005987.3 67 Nonsense Mutation CAG,TAG Q4* NP_005978.2

View Full Product Details