Product Details

SNP ID
rs199572964
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:222523655 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATCCGCTTACTCTTGGTTCTCAC[A/G]GGCACTGGCTTGTATTTGCACTTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HHIPL2 PubMed Links

Gene Details

Gene
HHIPL2
Gene Name
HHIP like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024746.3 1905 Silent Mutation NP_079022.2
XM_011509985.2 1905 Silent Mutation XP_011508287.1
XM_011509986.1 1905 Intron XP_011508288.1
XM_017002350.1 1905 Intron XP_016857839.1

View Full Product Details