Product Details

SNP ID
rs199561322
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:17375466 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATGGGCCCAACGAGGATGCCCCC[A/G]TGGGCACAGCTGTGCTGTACCTCAC
Phenotype
MIM: 610363
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PADI6 PubMed Links

Gene Details

Gene
PADI6
Gene Name
peptidyl arginine deiminase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207421.4 384 Missense Mutation ATG,GTG M112V NP_997304.3

View Full Product Details