Product Details

SNP ID
rs199599632
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:32077218 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCTCCATCCTGCTGTTCCTCACT[C/T]GCTTCACCGTTCTCACGGCAACAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR5585 PubMed Links

Gene Details

Gene
MIR5585
Gene Name
microRNA 5585
There are no transcripts associated with this gene.

Gene
TMEM39B
Gene Name
transmembrane protein 39B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319677.1 908 Missense Mutation CGC,TGC R37C NP_001306606.1
NM_001319678.1 908 Intron NP_001306607.1
NM_001319679.1 908 Intron NP_001306608.1
NM_018056.3 908 Missense Mutation CGC,TGC R164C NP_060526.2
XM_005270988.2 908 Missense Mutation CGC,TGC R37C XP_005271045.1
XM_006710724.1 908 Missense Mutation CGC,TGC R164C XP_006710787.1
XM_006710725.3 908 Missense Mutation CGC,TGC R164C XP_006710788.1
XM_011541681.2 908 Missense Mutation CGC,TGC R164C XP_011539983.1
XM_011541683.1 908 Missense Mutation CGC,TGC R37C XP_011539985.1
XM_011541684.2 908 Intron XP_011539986.1
XM_017001595.1 908 Missense Mutation CGC,TGC R49C XP_016857084.1

View Full Product Details