Product Details

SNP ID
rs199599943
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:85158829 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGTATCAATACTTTCTTGTAGA[A/T]CTTTGAGATTGTTTTTTCGATCCAC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SYDE2 PubMed Links

Gene Details

Gene
SYDE2
Gene Name
synapse defective Rho GTPase homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032184.1 2942 Missense Mutation GAT,GTT D1169V NP_115560.1
XM_005271254.3 2942 Intron XP_005271311.1
XM_006710946.3 2942 Intron XP_006711009.1
XM_017002483.1 2942 Intron XP_016857972.1
XM_017002484.1 2942 Intron XP_016857973.1
XM_017002485.1 2942 Missense Mutation GAT,GTT D911V XP_016857974.1
XM_017002486.1 2942 Missense Mutation GAT,GTT D911V XP_016857975.1

View Full Product Details