Product Details

SNP ID
rs199774410
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:154993684 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCCCCTCGAGACACTGGGCTGC[A/G]GGTGCCTGTCATTAAGATGGGCACA
Phenotype
MIM: 610595 MIM: 607377 MIM: 607646
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FLAD1 PubMed Links

Gene Details

Gene
FLAD1
Gene Name
flavin adenine dinucleotide synthetase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184891.1 99 Intron NP_001171820.1
NM_001184892.1 99 Intron NP_001171821.1
NM_025207.4 99 Intron NP_079483.3
NM_201398.2 99 Intron NP_958800.1
Gene
LENEP
Gene Name
lens epithelial protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018655.2 99 Missense Mutation CAG,CGG Q26R NP_061125.1
Gene
ZBTB7B
Gene Name
zinc finger and BTB domain containing 7B
There are no transcripts associated with this gene.

View Full Product Details