Product Details

SNP ID
rs199803522
Assay Type
Functionally Tested
NCBI dbSNP Submissions
12
Location
Chr.1:224356940 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGAGGAGGATGGAGGCGGTGGTG[G/T]TCGTCTTCTCTCTCCTCGATTGTTG
Phenotype
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
CNIH4 PubMed Links

Gene Details

Gene
CNIH4
Gene Name
cornichon family AMPA receptor auxiliary protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277197.1 130 Missense Mutation GTC,TTC V6F NP_001264126.1
NM_001277198.1 130 Missense Mutation GTC,TTC V6F NP_001264127.1
NM_001277199.1 130 UTR 5 NP_001264128.1
NM_001277200.1 130 Missense Mutation GTC,TTC V6F NP_001264129.1
NM_014184.3 130 Missense Mutation GTC,TTC V6F NP_054903.1

View Full Product Details