Product Details

SNP ID
rs199529688
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:203129138 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGAGCTCCATCCTGGCCCTGCT[C/G]GCAATTGCTGTGGACCGCTACCTCC
Phenotype
MIM: 102775
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADORA1 PubMed Links

Gene Details

Gene
ADORA1
Gene Name
adenosine A1 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000674.2 225 Silent Mutation CTC,CTG L99L NP_000665.1
NM_001048230.1 225 Silent Mutation CTC,CTG L99L NP_001041695.1
XM_005244901.1 225 UTR 5 XP_005244958.1
XM_005244902.4 225 UTR 5 XP_005244959.1
XM_017000342.1 225 UTR 5 XP_016855831.1

View Full Product Details