Product Details

SNP ID
rs200167027
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:36085214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTCGACTGGTGGAGAGGATTGAT[A/C]CTGTCAACCGGTGGAAGGAGATGCT
Phenotype
MIM: 610624 MIM: 608953
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADPRHL2 PubMed Links

Gene Details

Gene
ADPRHL2
Gene Name
ADP-ribosylhydrolase like 2
There are no transcripts associated with this gene.

Gene
TEKT2
Gene Name
tektin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014466.2 307 Missense Mutation ACT,CCT T70P NP_055281.2
XM_005270753.2 307 Missense Mutation ACT,CCT T70P XP_005270810.1
XM_011541258.2 307 Missense Mutation ACT,CCT T70P XP_011539560.1
XM_017001055.1 307 Missense Mutation ACT,CCT T70P XP_016856544.1

View Full Product Details