Product Details

SNP ID
rs200512361
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:246541200 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTCTCGCATCAAGTGGAGTCAAT[G/T]AACTGCAAAAGAAATACAAAGTAAA
Phenotype
MIM: 607055
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TFB2M PubMed Links

Gene Details

Gene
TFB2M
Gene Name
transcription factor B2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022366.2 Intron NP_071761.1
XM_011544248.2 Intron XP_011542550.1
XM_017002055.1 Intron XP_016857544.1

View Full Product Details