Product Details

SNP ID
rs200804642
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:39957179 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCTACCAGGTGACGGGCTGTGC[C/T]CTGGGTTTCTTCCTTCAGATCTACC
Phenotype
MIM: 614397
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MFSD2A PubMed Links

Gene Details

Gene
MFSD2A
Gene Name
major facilitator superfamily domain containing 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136493.2 334 Silent Mutation GCC,GCT A62A NP_001129965.1
NM_001287808.1 334 Intron NP_001274737.1
NM_001287809.1 334 Intron NP_001274738.1
NM_032793.4 334 Silent Mutation GCC,GCT A62A NP_116182.2
XM_005271285.1 334 Silent Mutation GCC,GCT A60A XP_005271342.1
XM_011542312.2 334 Silent Mutation GCC,GCT A62A XP_011540614.1

View Full Product Details