Product Details

SNP ID
rs200915100
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:152600898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGAAAGTGGCTGCTGCCTGAGCC[A/G]CCACAGGCACTTCAGGTCCCATCAA
Phenotype
MIM: 612615
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LCE3C PubMed Links

Gene Details

Gene
LCE3C
Gene Name
late cornified envelope 3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178434.2 Intron NP_848521.1

View Full Product Details