Product Details

SNP ID
rs200646741
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:26894373 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCCGGTCACGTCCTCATGCAGC[A/G]CTTCATGCCGTTCCCATTCCTCACC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPATCH3 PubMed Links

Gene Details

Gene
GPATCH3
Gene Name
G-patch domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022078.2 943 Missense Mutation GCG,GTG A305V NP_071361.2
Gene
GPN2
Gene Name
GPN-loop GTPase 2
There are no transcripts associated with this gene.

View Full Product Details