Product Details
- SNP ID
-
rs200328355
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:225988262 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACTGGGAAATTTGGCTGCCATCTC[A/G]ACACCATTGCTACCAATTTTTGGAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SDE2
PubMed Links
Gene Details
- Gene
- SDE2
- Gene Name
- SDE2 telomere maintenance homolog
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152608.3 |
821 |
Silent Mutation |
GTC,GTT |
V256V |
NP_689821.3 |
View Full Product Details