Product Details

SNP ID
rs200872114
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:161984078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCACAGCAAAGTCCACGTCTGAG[C/T]GGCCCTGCCATCGCCAGGGGGTGGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OLFML2B PubMed Links

Gene Details

Gene
OLFML2B
Gene Name
olfactomedin like 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297713.1 2891 Missense Mutation CAC,CGC H618R NP_001284642.1
NM_015441.2 2891 Missense Mutation CAC,CGC H617R NP_056256.1
XM_005245075.2 2891 Missense Mutation CAC,CGC H619R XP_005245132.1
XM_011509398.2 2891 Missense Mutation CAC,CGC H377R XP_011507700.1
XM_017000967.1 2891 Intron XP_016856456.1

View Full Product Details