Product Details

SNP ID
rs200636744
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:53894534 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCAGGACAGAGGTGCAACATTTG[A/G]GAGTTTATGCAAGGTCAGGAGGCTG
Phenotype
MIM: 147892
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DIO1 PubMed Links

Gene Details

Gene
DIO1
Gene Name
deiodinase, iodothyronine type I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000792.6 348 Nonsense Mutation TGA,TGG *108W NP_000783.2
NM_001039715.2 348 Nonsense Mutation TGA,TGG *108W NP_001034804.1
NM_001039716.2 348 Nonsense Mutation TGA,TGG *108W NP_001034805.1
NM_001324316.1 348 Nonsense Mutation TGA,TGG *108W NP_001311245.1
NM_213593.4 348 Intron NP_998758.1
Gene
YIPF1
Gene Name
Yip1 domain family member 1
There are no transcripts associated with this gene.

View Full Product Details