Product Details

SNP ID
rs200796048
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:152972071 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAATGCCCACCGAAAGGCACCATC[A/G]TTCCAGCCCAGCAGAAGTGTCCCTC
Phenotype
MIM: 616363
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPRR4 PubMed Links

Gene Details

Gene
SPRR4
Gene Name
small proline rich protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173080.2 939 Missense Mutation ATT,GTT I61V NP_775103.1
XM_017000482.1 939 Missense Mutation ATT,GTT I61V XP_016855971.1

View Full Product Details