Product Details

SNP ID
rs200185591
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:197905833 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTATGTGGATCCAGCTACTGGCT[A/G]TGTGGTGCTCACACAGATTGCCCAC
Phenotype
MIM: 606066
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf53 PubMed Links

Gene Details

Gene
C1orf53
Gene Name
chromosome 1 open reading frame 53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024594.2 400 Missense Mutation TAT,TGT Y101C NP_001019765.1
Gene
LHX9
Gene Name
LIM homeobox 9
There are no transcripts associated with this gene.

View Full Product Details