Product Details

SNP ID
rs200213184
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:162381951 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACTCCAAAGCTCCAGGCCAGCC[A/G]CTCCTTCCCCCACTTGTCCAAGCCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf111 PubMed Links

Gene Details

Gene
C1orf111
Gene Name
chromosome 1 open reading frame 111
There are no transcripts associated with this gene.

Gene
C1orf226
Gene Name
chromosome 1 open reading frame 226
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085375.1 222 Missense Mutation CAC,CGC H17R NP_001078844.1
NM_001135240.1 222 Missense Mutation CAC,CGC H60R NP_001128712.1

View Full Product Details